Learn More
Biomatik Corporation Recombinant Parathyroid Hormone (PTH), 100UG
Supplier:  Biomatik Corporation RPU43181
Encompass_Preferred
Recombinant Mouse Parathyroid Hormone (PTH) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Parathyroid Hormone (PTH). Target Synonyms: iPTH; Intact Parathyroid Hormone; Parathormone; Parathyrin. Accession Number: Q9Z0L6. Gene ID: 19226. Expression Region: Lys26~Gln115. Tag Info: N-terminal His and GST Tag. Theoretical MW: 40kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5692