Learn More
Biomatik Corporation Recombinant Parathyroid Hormone (PTH), 100UG
Catalog No. 501505693
Encompass_Preferred
Recombinant Rat Parathyroid Hormone (PTH) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Rat (Rattus norvegicus). Target Name: Parathyroid Hormone (PTH). Target Synonyms: iPTH; Intact Parathyroid Hormone; Parathormone; Parathyrin. Accession Number: P04089. Gene ID: 24694. Expression Region: Ala32~Gln115. Tag Info: Two N-terminal Tags, His-tag and MBP-tag. Theoretical MW: 59kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5693 Supplier Biomatik Corporation Supplier No. RPU43183
Recombinant Rat Parathyroid Hormone (PTH) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Rat (Rattus norvegicus). Target Name: Parathyroid Hormone (PTH). Target Synonyms: iPTH; Intact Parathyroid Hormone; Parathormone; Parathyrin. Accession Number: P04089. Gene ID: 24694. Expression Region: Ala32~Gln115. Tag Info: Two N-terminal Tags, His-tag and MBP-tag. Theoretical MW: 59kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.