Learn More
Biomatik Corporation Recombinant Parathyroid Hormone Related Protein (PTHrP), 100UG
Supplier:  Biomatik Corporation RPU43185
Encompass_Preferred
Recombinant Mouse Parathyroid Hormone Related Protein (PTHrP) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Parathyroid Hormone Related Protein (PTHrP). Target Synonyms: PTHLH; HHM; PTHR; PLP; Osteostatin; Parathyroid Hormone Like Hormone. Accession Number: P22858. Expression Region: Ala37~His175. Tag Info: N-terminal His Tag. Theoretical MW: 18/19kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5694