Learn More
Biomatik Corporation Recombinant Peripheral Myelin Protein 22 (PMP22), 100UG
Catalog No. 501505697
Encompass_Preferred
Recombinant Human Peripheral Myelin Protein 22 (PMP22) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Peripheral Myelin Protein 22 (PMP22). Target Synonyms: DSS; CMT1A; CMT1E; GAS3; HMSNIA; HNPP; Sp110; Growth arrest-specific protein 3. Accession Number: Q01453. Expression Region: Gly31~Gly133. Tag Info: Two N-terminal Tags, His-tag and SUMO-tag. Theoretical MW: 22/19kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5697 Supplier Biomatik Corporation Supplier No. RPU43222
Recombinant Human Peripheral Myelin Protein 22 (PMP22) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Peripheral Myelin Protein 22 (PMP22). Target Synonyms: DSS; CMT1A; CMT1E; GAS3; HMSNIA; HNPP; Sp110; Growth arrest-specific protein 3. Accession Number: Q01453. Expression Region: Gly31~Gly133. Tag Info: Two N-terminal Tags, His-tag and SUMO-tag. Theoretical MW: 22/19kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.