Learn More
Biomatik Corporation Recombinant Phospholipase A2, Cytosolic (cPLA2), 100UG
Supplier:  Biomatik Corporation RPU43288
Encompass_Preferred
Recombinant Human Cytosolic Phospholipase A2 (PLA2G4) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Cytosolic Phospholipase A2 (PLA2G4). Target Synonyms: c-PLA2; cPLA2; PLA2G4A; Phospholipase A2, Cytosolic; Phospholipase A2,Group IV(Cytosolic,Calcium-Dependent); Phosphatidylcholine 2-acylhydrolase; Lysophospholipase. Accession Number: P47712. Gene ID: 5321. Expression Region: Met1~Ser178. Tag Info: N-terminal His Tag. Theoretical MW: 26kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5699