Learn More
Biomatik Corporation Recombinant Placenta Growth Factor (PLGF), 100UG
Supplier:  Biomatik Corporation RPU43307
Encompass_Preferred
Recombinant Human Placenta Growth Factor (PLGF) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Placenta Growth Factor (PLGF). Target Synonyms: PlGF2; PGF; PGFL; Placental Growth Factor-Like; Vascular Endothelial Growth Factor-Related Protein. Accession Number: P49763. Gene ID: 5228. Expression Region: Phe43~Leu162. Tag Info: N-terminal His Tag. Theoretical MW: 18kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5701