Learn More
Biomatik Corporation Recombinant Procalcitonin (PCT), 100UG
Supplier:  Biomatik Corporation RPU43415
Encompass_Preferred
Recombinant Human Procalcitonin (PCT) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Procalcitonin (PCT). Target Synonyms: Pro-Calcitonin. Accession Number: P01258. Gene ID: 796. Expression Region: Ala26~Asn141. Tag Info: N-terminal His Tag. Theoretical MW: 17kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5704