Learn More
Biomatik Corporation Recombinant Programmed Cell Death Protein 1 Ligand 1 (PDCD1LG1), 100UG
Supplier:  Biomatik Corporation RPU43427
Encompass_Preferred
Recombinant Human Programmed Cell Death Protein 1 Ligand 1 (PDL1) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Programmed Cell Death Protein 1 Ligand 1 (PDL1). Target Synonyms: CD274; PDCD1LG1; B7-H; B7H1; PD-L1; PDCD1L1; PDCD1-LG1; Programed Death Ligand 1. Accession Number: Q9NZQ7. Gene ID: 29126. Expression Region: Pro24~Pro234. Tag Info: N-terminal His Tag. Theoretical MW: 32kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5705