Learn More
Biomatik Corporation Recombinant Prostaglandin E Synthase, Microsomal (PTGES), 100UG
Supplier:  Biomatik Corporation RPU43446
Encompass_Preferred
Recombinant Human Prostaglandin E Synthase, Microsomal (PTGES) is a purified Prokaryotic Protein. Purity: >80% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Prostaglandin E Synthase, Microsomal (PTGES). Target Synonyms: MGST1L1; MGST-IV; MGST1-L1; PGES; PIG12; PP102; PP1294; TP53I12; PGE Synthase; Tumor Protein p53 Inducible Protein 12; Glutathione S-Transferase 1-Like 1; MGST1-Like 1. Accession Number: O14684. Gene ID: 9536. Expression Region: Thr21~Ala148. Tag Info: N-terminal His and GST Tag. Theoretical MW: 44/33/28kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5706