Learn More
Biomatik Corporation Recombinant Prostaglandin Endoperoxide Synthase 2 (PTGS2), 100UG
Supplier:  Biomatik Corporation RPU43448
Encompass_Preferred
Recombinant Mouse Cyclooxygenase-2 (COX 2) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Cyclooxygenase-2 (COX 2). Target Synonyms: PGG/HS; PGHS2; PHS2; HCox2; PTGS2; Prostaglandin Endoperoxide Synthase 2; Prostaglandin G/H Synthase And Cyclooxygenase; Prostaglandin H2 synthase 2. Accession Number: Q05769. Gene ID: 19225. Expression Region: Tyr240~Leu351. Tag Info: N-terminal His Tag. Theoretical MW: 14kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5707