Learn More
Biomatik Corporation Recombinant Human 78KDA glucose-regulated protein (HSPA5), partial, 100UG
Catalog No. 501504647
Encompass_Preferred
Recombinant Human Endoplasmic reticulum chaperone BiP (HSPA5), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: HSPA5. Target Synonyms: Endoplasmic reticulum lumenal Ca(2+)-binding protein grp78Heat shock 70 kDa protein 5Immunoglobulin heavy chain-binding protein; BiP. Accession Number: P11021. Expression Region: 25~293aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 33.6kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4647 Supplier Biomatik Corporation Supplier No. RPC20182
Recombinant Human Endoplasmic reticulum chaperone BiP (HSPA5), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: HSPA5. Target Synonyms: Endoplasmic reticulum lumenal Ca(2+)-binding protein grp78Heat shock 70 kDa protein 5Immunoglobulin heavy chain-binding protein; BiP. Accession Number: P11021. Expression Region: 25~293aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 33.6kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.