Learn More
Biomatik Corporation Recombinant Prostate Stem Cell Antigen (PSCA), 100UG
Supplier:  Biomatik Corporation RPU43450
Encompass_Preferred
Recombinant Human Prostate Stem Cell Antigen (PSCA) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Prostate Stem Cell Antigen (PSCA). Accession Number: O43653. Gene ID: 8000. Expression Region: Leu13~Ser86. Tag Info: N-terminal His Tag. Theoretical MW: 13kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5709