Learn More
Biomatik Corporation Recombinant Proteinase 3 (PR3), 100UG
Catalog No. 501505712
Encompass_Preferred
Recombinant Rat Proteinase 3 (PR3) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Rat (Rattus norvegicus). Target Name: Proteinase 3 (PR3). Target Synonyms: MBN; PRTN3; ACPA; AGP7; C-ANCA; MBT; P29; PRK; Myeloblastin; Serine Proteinase,Neutrophil,Wegener Granulomatosis Autoantigen. Accession Number: Q8K597. Gene ID: 314615. Expression Region: Ala18~His246. Tag Info: N-terminal His Tag. Theoretical MW: 26kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5712 Supplier Biomatik Corporation Supplier No. RPU43520
Recombinant Rat Proteinase 3 (PR3) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Rat (Rattus norvegicus). Target Name: Proteinase 3 (PR3). Target Synonyms: MBN; PRTN3; ACPA; AGP7; C-ANCA; MBT; P29; PRK; Myeloblastin; Serine Proteinase,Neutrophil,Wegener Granulomatosis Autoantigen. Accession Number: Q8K597. Gene ID: 314615. Expression Region: Ala18~His246. Tag Info: N-terminal His Tag. Theoretical MW: 26kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.