Learn More
Biomatik Corporation Recombinant RAR Related Orphan Receptor Gamma (RORg), 100UG
Supplier:  Biomatik Corporation RPU43560
Encompass_Preferred
Recombinant Human RAR Related Orphan Receptor Gamma (RORg) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: RAR Related Orphan Receptor Gamma (RORg). Target Synonyms: RORC; NR1F3; RORG; RZRG; TOR; Nuclear receptor subfamily 1 group F member 3; RAR-related orphan receptor C; Retinoid-related orphan receptor-gamma. Accession Number: P51449. Gene ID: 6097. Expression Region: Glu212~Ser461. Tag Info: N-terminal His Tag. Theoretical MW: 32kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5714