Learn More
Biomatik Corporation Recombinant Retinol Binding Protein 4, Plasma (RBP4), 100UG
Supplier:  Biomatik Corporation RPU43621
Encompass_Preferred
Recombinant Bovine Retinol Binding Protein 4 (RBP4) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Bovine (Bos taurus). Target Name: Retinol Binding Protein 4 (RBP4). Target Synonyms: PRBP; RBP; Plasma retinol-binding protein; Retinol Binding Protein 4, Plasma. Accession Number: P18902. Expression Region: Glu1~Leu183. Tag Info: N-terminal His Tag. Theoretical MW: 22kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5717