Learn More
Biomatik Corporation Recombinant Ribonuclease A3 (RNASE3), 100UG
Catalog No. 501505718
Encompass_Preferred
Recombinant Human Ribonuclease A3 (RNASE3) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Ribonuclease A3 (RNASE3). Target Synonyms: Rnase-A3; RnaseA3; ECP; RNS3; Ribonuclease,RNase A Family 3; Eosinophil Cationic Protein. Accession Number: P12724. Gene ID: 6037. Expression Region: Pro30~Val154. Tag Info: N-terminal His Tag. Theoretical MW: 19kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5718 Supplier Biomatik Corporation Supplier No. RPU43643
Recombinant Human Ribonuclease A3 (RNASE3) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Ribonuclease A3 (RNASE3). Target Synonyms: Rnase-A3; RnaseA3; ECP; RNS3; Ribonuclease,RNase A Family 3; Eosinophil Cationic Protein. Accession Number: P12724. Gene ID: 6037. Expression Region: Pro30~Val154. Tag Info: N-terminal His Tag. Theoretical MW: 19kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.