Learn More
Biomatik Corporation Recombinant S100 Calcium Binding Protein A8 (S100A8), 100UG
Supplier:  Biomatik Corporation RPU43698
Encompass_Preferred
Recombinant Human S100 Calcium Binding Protein A8 (S100A8) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: S100 Calcium Binding Protein A8 (S100A8). Target Synonyms: MRP8; MIF; 60B8AG; CAGA; CFAG; CGLA; CP-10; L1Ag; NIF; P8; Calgranulin A; Cystic fibrosis antigen; Leukocyte L1 complex light chain; Migration inhibitory factor-related protein 8. Accession Number: P05109. Gene ID: 6279. Expression Region: Met1~Glu93. Tag Info: N-terminal His Tag. Theoretical MW: 12kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5720