Learn More
Biomatik Corporation Recombinant S100 Calcium Binding Protein B (S100B), 100UG
Supplier:  Biomatik Corporation RPU43704
Encompass_Preferred
Recombinant Mouse S100 Calcium Binding Protein B (S100B) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: S100 Calcium Binding Protein B (S100B). Target Synonyms: S100-B; NEF; S100Beta; S-100 protein subunit beta. Accession Number: P50114. Gene ID: 20203. Expression Region: Met1~Glu92. Tag Info: N-terminal His Tag. Theoretical MW: 10kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5722