Learn More
Biomatik Corporation Recombinant Secretogranin II (SCG2), 100UG
Supplier:  Biomatik Corporation RPU43726
Encompass_Preferred
Recombinant Mouse Secretogranin II (SCG2) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Secretogranin II (SCG2). Target Synonyms: SN; SgII; CHGC; Chromogranin C; Secretoneurin. Accession Number: Q03517. Gene ID: 20254. Expression Region: Gln342~Leu609. Tag Info: N-terminal His Tag. Theoretical MW: 39kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5723