Learn More
Biomatik Corporation Recombinant Secretory Leukocyte Peptidase Inhibitor (SLPI), 100UG
Supplier:  Biomatik Corporation RPU43729
Encompass_Preferred
Recombinant Mouse Secretory Leukocyte Peptidase Inhibitor (SLPI) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Secretory Leukocyte Peptidase Inhibitor (SLPI). Target Synonyms: ALK1; ALP; MPI; BLPI; HUSI; HUSI-I; WAP4; WFDC4; Antileukoproteinase; Seminal proteinase inhibitor; Mucus proteinase inhibitor; WAP four-disulfide core domain protein 4. Accession Number: P97430. Gene ID: 20568. Expression Region: Pro20~Met131. Tag Info: N-terminal His Tag. Theoretical MW: 20kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5724