Learn More
Biomatik Corporation Recombinant Sialic Acid Binding Ig Like Lectin 1 (SIGLEC1), 100UG
Catalog No. 501505727
Encompass_Preferred
Recombinant Human Sialoadhesin (SN) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Sialoadhesin (SN). Target Synonyms: CD169; SIGLEC1; Sialic Acid Binding Ig Like Lectin 1. Accession Number: Q9BZZ2. Gene ID: 6614. Expression Region: Pro1272~Leu1411. Tag Info: N-terminal His Tag. Theoretical MW: 18kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5727 Supplier Biomatik Corporation Supplier No. RPU43782
Recombinant Human Sialoadhesin (SN) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Sialoadhesin (SN). Target Synonyms: CD169; SIGLEC1; Sialic Acid Binding Ig Like Lectin 1. Accession Number: Q9BZZ2. Gene ID: 6614. Expression Region: Pro1272~Leu1411. Tag Info: N-terminal His Tag. Theoretical MW: 18kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.