Learn More
Biomatik Corporation Recombinant Slit Homolog 2 (Slit2), 100UG
Catalog No. 501505728
Encompass_Preferred
Recombinant Rat Slit Homolog 2 (Slit2) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Rat (Rattus norvegicus). Target Name: Slit Homolog 2 (Slit2). Target Synonyms: SLIL3. Accession Number: Q9WVC1. Expression Region: Asn209~Gly374. Tag Info: N-terminal His Tag. Theoretical MW: 20kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5728 Supplier Biomatik Corporation Supplier No. RPU43842
Recombinant Rat Slit Homolog 2 (Slit2) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Rat (Rattus norvegicus). Target Name: Slit Homolog 2 (Slit2). Target Synonyms: SLIL3. Accession Number: Q9WVC1. Expression Region: Asn209~Gly374. Tag Info: N-terminal His Tag. Theoretical MW: 20kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.