Learn More
Biomatik Corporation Recombinant Solute Carrier Family 30 Member 8 (SLC30A8), 100UG
Supplier:  Biomatik Corporation RPU43855
Encompass_Preferred
Recombinant Human Solute Carrier Family 30 Member 8 (SLC30A8) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Solute Carrier Family 30 Member 8 (SLC30A8). Target Synonyms: ZNT8; Zinc Transporter 8. Accession Number: Q8IWU4. Gene ID: 169026. Expression Region: Met1~Ile80. Tag Info: N-terminal His Tag. Theoretical MW: 10/15kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5730