Learn More
Biomatik Corporation Recombinant Somatostatin (SST), 100UG
Supplier:  Biomatik Corporation RPU43856
Encompass_Preferred
Recombinant Human Somatostatin (SST) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Somatostatin (SST). Target Synonyms: GHIH; SRIF; SMST; GHRIH; GHRIF; Growth Hormone Releasing Inhibiting Hormone; Somatotropin Release-Inhibiting Factor; Somatostatin-14; Somatostatin-28. Accession Number: P61278. Gene ID: 6750. Expression Region: Met1~Cys116. Tag Info: N-terminal His Tag. Theoretical MW: 18/20kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5731