Learn More
Biomatik Corporation Recombinant Stromal Cell Derived Factor 1 (SDF1), 100UG
Supplier:  Biomatik Corporation RPU43895
Encompass_Preferred
Recombinant Mouse Stromal Cell Derived Factor 1 (SDF1) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Stromal Cell Derived Factor 1 (SDF1). Target Synonyms: CXCL12; PBSF; P-BSF; SCYB12; SDF1-A; SDF1-B; SDF1A; SDF1B; TLSF-A; TLSF-B; TPAR1; Pre-B Cell Growth-Stimulating Factor; Chemokine C-X-C-Motif Ligand 12. Accession Number: P40224. Gene ID: 20315. Expression Region: Lys22~Lys89. Tag Info: N-terminal His Tag. Theoretical MW: 11kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5734