Learn More
Biomatik Corporation Recombinant Surfactant Associated Protein D (SPD), 100UG
Supplier:  Biomatik Corporation RPU43931
Encompass_Preferred
Recombinant Human Surfactant Associated Protein D (SPD) is a purified Prokaryotic Protein. Purity: >80% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Surfactant Associated Protein D (SPD). Target Synonyms: SFTPD; COLEC7; PSP-D; SFTP4; SP-D; Pulmonary Surfactant Protein D; Collectin-7; Lung surfactant protein D. Accession Number: P35247. Gene ID: 6441. Expression Region: Ala21~Phe375. Tag Info: N-terminal His Tag. Theoretical MW: 43kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5736