Learn More
Biomatik Corporation Recombinant T-Cell Surface Glycoprotein CD3 Epsilon (CD3e), 100UG
Catalog No. 501505739
Encompass_Preferred
Recombinant Human T-Cell Surface Glycoprotein CD3 Epsilon (CD3e) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: T-Cell Surface Glycoprotein CD3 Epsilon (CD3e). Target Synonyms: T3E; TCRE; CD3-E Molecule,Epsilon(CD3-TCR Complex); T-cell surface antigen T3/Leu-4 epsilon chain. Accession Number: P07766. Gene ID: 916. Expression Region: Met28~Asn185. Tag Info: N-terminal His Tag. Theoretical MW: 22kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5739 Supplier Biomatik Corporation Supplier No. RPU43962
Recombinant Human T-Cell Surface Glycoprotein CD3 Epsilon (CD3e) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: T-Cell Surface Glycoprotein CD3 Epsilon (CD3e). Target Synonyms: T3E; TCRE; CD3-E Molecule,Epsilon(CD3-TCR Complex); T-cell surface antigen T3/Leu-4 epsilon chain. Accession Number: P07766. Gene ID: 916. Expression Region: Met28~Asn185. Tag Info: N-terminal His Tag. Theoretical MW: 22kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.