Learn More
Biomatik Corporation Recombinant Telomerase Reverse Transcriptase (TERT), 100UG
Supplier:  Biomatik Corporation RPU43964
Encompass_Preferred
Recombinant Mouse Telomerase Reverse Transcriptase (TERT) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Telomerase Reverse Transcriptase (TERT). Target Synonyms: EST2; TCS1; TP2; TRT; hEST2; Telomerase catalytic subunit; Telomerase-associated protein 2. Accession Number: O70372. Gene ID: 21752. Expression Region: Gln642~Lys974. Tag Info: N-terminal His and GST Tag. Theoretical MW: 60kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5740