Learn More
Biomatik Corporation Recombinant Thymic Stromal Lymphopoietin (TSLP), 100UG
Supplier:  Biomatik Corporation RPU44001
Encompass_Preferred
Recombinant Rat Thymic Stromal Lymphopoietin (TSLP) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Rat (Rattus norvegicus). Target Name: Thymic Stromal Lymphopoietin (TSLP). Accession Number: A0A0G2JUJ3. Expression Region: Leu75~Ser196. Tag Info: N-terminal His Tag. Theoretical MW: 15kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5741