Learn More
Biomatik Corporation Recombinant Thymosin Beta 4 (Tb4), 100UG
Catalog No. 501505742
Encompass_Preferred
Recombinant Human Thymosin Beta 4 (TMSB4X) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Thymosin Beta 4 (TMSB4X). Target Synonyms: Tb4; FX; PTMB4; TB4X; TMSB4; Thymosin,Beta 4,X-Linked; TB-500; Seraspenide; Hematopoietic system regulatory peptide. Accession Number: P62328. Gene ID: 7114. Expression Region: Met1~Ser44. Tag Info: N-terminal His Tag. Theoretical MW: 6kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5742 Supplier Biomatik Corporation Supplier No. RPU44009
Recombinant Human Thymosin Beta 4 (TMSB4X) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Thymosin Beta 4 (TMSB4X). Target Synonyms: Tb4; FX; PTMB4; TB4X; TMSB4; Thymosin,Beta 4,X-Linked; TB-500; Seraspenide; Hematopoietic system regulatory peptide. Accession Number: P62328. Gene ID: 7114. Expression Region: Met1~Ser44. Tag Info: N-terminal His Tag. Theoretical MW: 6kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.