Learn More
Biomatik Corporation Recombinant Tissue Factor (TF), 100UG
Supplier:  Biomatik Corporation RPU44024
Encompass_Preferred
Recombinant Human Tissue Factor (TF) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Tissue Factor (TF). Target Synonyms: CD142; FIII; F3; TFA; Thromboplastin; Coagulation Factor III. Accession Number: P13726. Gene ID: 2152. Expression Region: Gly34~glu251. Tag Info: N-terminal His Tag. Theoretical MW: 33kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5744