Learn More
Biomatik Corporation Recombinant Tissue Factor (TF), 100UG
Supplier:  Biomatik Corporation RPU44026
Encompass_Preferred
Recombinant Rat Tissue Factor (TF) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Rat (Rattus norvegicus). Target Name: Tissue Factor (TF). Target Synonyms: CD142; FIII; F3; TFA; Thromboplastin; Coagulation Factor III. Accession Number: P42533. Expression Region: Gly30~Glu252. Tag Info: N-terminal His Tag. Theoretical MW: 29kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5745