Learn More
Biomatik Corporation Recombinant Toll Like Receptor 4 (TLR4), 100UG
Supplier:  Biomatik Corporation RPU44074
Encompass_Preferred
Recombinant Mouse Toll Like Receptor 4 (TLR4) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Toll Like Receptor 4 (TLR4). Target Synonyms: CD284; TOLL; HToll. Accession Number: Q9QUK6. Gene ID: 21898. Expression Region: Asn26~Glu269. Tag Info: N-terminal His Tag. Theoretical MW: 38kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5747