Learn More
Biomatik Corporation Recombinant Transferrin Receptor 2 (TFR2), 100UG
Supplier: Biomatik Corporation RPU44120
Recombinant Mouse Transferrin Receptor 2 (TFR2) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Transferrin Receptor 2 (TFR2). Target Synonyms: HFE3; TFRC2. Accession Number: Q9JKX3. Gene ID: 50765. Expression Region: Ile86~Val259. Tag Info: N-terminal His Tag. Theoretical MW: 29kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.