Learn More
Biomatik Corporation Recombinant Transforming Growth Factor Beta 1 (TGFb1), 100UG
Catalog No. 501505753
Encompass_Preferred
Recombinant Human Transforming Growth Factor Beta 1 (TGFb1) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Transforming Growth Factor Beta 1 (TGFb1). Target Synonyms: TGF-B1; CED; DPD1; LAP; Camurati-Engelmann Disease; Latency-associated peptide. Accession Number: P01137. Gene ID: 7040. Expression Region: Ala279~Ser390. Tag Info: N-terminal His Tag. Theoretical MW: 15kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5753 Supplier Biomatik Corporation Supplier No. RPU44129
Recombinant Human Transforming Growth Factor Beta 1 (TGFb1) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Transforming Growth Factor Beta 1 (TGFb1). Target Synonyms: TGF-B1; CED; DPD1; LAP; Camurati-Engelmann Disease; Latency-associated peptide. Accession Number: P01137. Gene ID: 7040. Expression Region: Ala279~Ser390. Tag Info: N-terminal His Tag. Theoretical MW: 15kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.