Learn More
Biomatik Corporation Recombinant Transglutaminase 3, Epidermal (TGM3), 100UG
Supplier:  Biomatik Corporation RPU44148
Encompass_Preferred
Recombinant Mouse Transglutaminase 3, Epidermal (TGM3) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Transglutaminase 3, Epidermal (TGM3). Target Synonyms: TGE; Egt; Protein-Glutamine Gamma-Glutamyltransferase E; Transglutaminase E. Accession Number: Q08189. Gene ID: 21818. Expression Region: Ala468~Glu693. Tag Info: N-terminal His Tag. Theoretical MW: 35kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5756