Learn More
Biomatik Corporation Recombinant Trefoil Factor 3, Intestinal (TFF3), 100UG
Supplier:  Biomatik Corporation RPU44171
Encompass_Preferred
Recombinant Dog Trefoil Factor 3 (TFF3) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Dog (Canis familiaris; Canine). Target Name: Trefoil Factor 3 (TFF3). Target Synonyms: HITF; ITF3; TFI; HP1.B; Trefoil Factor 3, Intestinal. Accession Number: Q863B4. Gene ID: 403488. Expression Region: Tyr24~Phe80. Tag Info: N-terminal His Tag. Theoretical MW: 13kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5760