Learn More
Biomatik Corporation Recombinant Troponin T Type 1, Slow Skeletal (TNNT1), 100UG
Catalog No. 501505763
Encompass_Preferred
Recombinant Human Troponin T Type 1, Slow Skeletal (TNNT1) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Troponin T Type 1, Slow Skeletal (TNNT1). Target Synonyms: ANM; STNT; TNT; TNTS; Slow Skeletal Muscle Troponin T. Accession Number: P13805. Gene ID: 7138. Expression Region: Met1~Gly259. Tag Info: N-terminal His Tag. Theoretical MW: 40kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5763 Supplier Biomatik Corporation Supplier No. RPU44191
Recombinant Human Troponin T Type 1, Slow Skeletal (TNNT1) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Troponin T Type 1, Slow Skeletal (TNNT1). Target Synonyms: ANM; STNT; TNT; TNTS; Slow Skeletal Muscle Troponin T. Accession Number: P13805. Gene ID: 7138. Expression Region: Met1~Gly259. Tag Info: N-terminal His Tag. Theoretical MW: 40kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.