Learn More
Biomatik Corporation Recombinant Tryptase (TPS), 100UG
Supplier:  Biomatik Corporation RPU44196
Encompass_Preferred
Recombinant Dog Tryptase (TPS) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Dog (Canis familiaris; Canine). Target Name: Tryptase (TPS). Target Synonyms: TPSAB1; Tryptase Alpha/Beta 1; Tryptase Alpha II; Tryptase Beta I; Tryptase-I; Tryptase-II; Tryptase-III. Accession Number: P15944. Gene ID: 100049001. Expression Region: Thr151~Pro275. Tag Info: N-terminal His Tag. Theoretical MW: 14kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5764