Learn More
Biomatik Corporation Recombinant Tumor Necrosis Factor Alpha (TNFa), 100UG
Catalog No. 501505767
Encompass_Preferred
Recombinant Rat Tumor Necrosis Factor Alpha (TNFa) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Rat (Rattus norvegicus). Target Name: Tumor Necrosis Factor Alpha (TNFa). Target Synonyms: DIF; TNF-A; TNFSF2; Cachectin; Tumor Necrosis Factor Ligand Superfamily Member 2. Accession Number: P16599. Gene ID: 24835. Expression Region: Leu80~Leu235. Tag Info: N-terminal His Tag. Theoretical MW: 21kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5767 Supplier Biomatik Corporation Supplier No. RPU44220
Recombinant Rat Tumor Necrosis Factor Alpha (TNFa) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Rat (Rattus norvegicus). Target Name: Tumor Necrosis Factor Alpha (TNFa). Target Synonyms: DIF; TNF-A; TNFSF2; Cachectin; Tumor Necrosis Factor Ligand Superfamily Member 2. Accession Number: P16599. Gene ID: 24835. Expression Region: Leu80~Leu235. Tag Info: N-terminal His Tag. Theoretical MW: 21kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.