Learn More
Biomatik Corporation Recombinant Tumor Necrosis Factor Beta (TNFb), 100UG
Supplier:  Biomatik Corporation RPU44229
Encompass_Preferred
Recombinant Mouse Tumor Necrosis Factor Beta (TNFb) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Tumor Necrosis Factor Beta (TNFb). Target Synonyms: LT; LTA; TNF-B; TNFSF1; Tumor Necrosis Factor Ligand Superfamily Member 1; Lymphotoxin Alpha. Accession Number: P09225. Gene ID: 16992. Expression Region: Pro49~Leu202. Tag Info: N-terminal His Tag. Theoretical MW: 17kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5768