Learn More
Biomatik Corporation Recombinant Escherichia coli 30S ribosomal protein S4 (rpsD), 100UG
Supplier:  Biomatik Corporation RPC20190
Encompass_Preferred
Recombinant Escherichia coli 30S ribosomal protein S4 (rpsD) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Escherichia coli (strain K12). Target Name: rpsD. Target Synonyms: rpsD; ramA; b3296; JW3258; 30S ribosomal protein S4; Small ribosomal subunit protein uS4. Accession Number: P0A7V8. Expression Region: 2~206aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 27.3kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4653