Learn More
Biomatik Corporation Recombinant Vascular Endothelial Growth Factor Receptor 1 (VEGFR1), 100UG
Catalog No. 501505778
Encompass_Preferred
Recombinant Rat Vascular Endothelial Growth Factor Receptor 1 (VEGFR1) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Rat (Rattus norvegicus). Target Name: Vascular Endothelial Growth Factor Receptor 1 (VEGFR1). Target Synonyms: Flt1; Fms-Related Tyrosine Kinase 1; Vascular Permeability Factor Receptor; Fms-like tyrosine kinase 1; Tyrosine-protein kinase receptor FLT. Accession Number: P53767. Expression Region: Ser303~Val524. Tag Info: N-terminal His Tag. Theoretical MW: 32kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5778 Supplier Biomatik Corporation Supplier No. RPU44331
Recombinant Rat Vascular Endothelial Growth Factor Receptor 1 (VEGFR1) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Rat (Rattus norvegicus). Target Name: Vascular Endothelial Growth Factor Receptor 1 (VEGFR1). Target Synonyms: Flt1; Fms-Related Tyrosine Kinase 1; Vascular Permeability Factor Receptor; Fms-like tyrosine kinase 1; Tyrosine-protein kinase receptor FLT. Accession Number: P53767. Expression Region: Ser303~Val524. Tag Info: N-terminal His Tag. Theoretical MW: 32kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.