Learn More
Biomatik Corporation Recombinant Vascular Endothelial Growth Factor Receptor 2 (VEGFR2), 100UG
Supplier:  Biomatik Corporation RPU44334
Encompass_Preferred
Recombinant Human Vascular Endothelial Growth Factor Receptor 2 (VEGFR2) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Vascular Endothelial Growth Factor Receptor 2 (VEGFR2). Target Synonyms: CD309; FLK1; VEGFR; KDR; A Type III Receptor Tyrosine Kinase; Kinase Insert Domain Receptor; Kinase Insert Domain Receptor; Fetal Liver Kinase-1. Accession Number: P35968. Gene ID: 3791. Expression Region: Asn46~Thr320. Tag Info: N-terminal His Tag. Theoretical MW: 33kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5780