Learn More
Biomatik Corporation Recombinant Versican (VS), 100UG
Supplier:  Biomatik Corporation RPU44344
Encompass_Preferred
Recombinant Human Versican (VCAN) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Versican (VCAN). Target Synonyms: CSPG2; VS; ERVR; PG-M; WGN1; Chondroitin Sulfate Proteoglycan 2; Chondroitin sulfate proteoglycan core protein 2; Glial hyaluronate-binding; Large fibroblast proteoglycan. Accession Number: P13611. Gene ID: 1462. Expression Region: Thr2958~Ser3209. Tag Info: N-terminal His Tag. Theoretical MW: 38kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5781