Learn More
Biomatik Corporation Recombinant Von Willebrand Factor (vWF), 100UG
Supplier:  Biomatik Corporation RPU44368
Encompass_Preferred
Recombinant Mouse Von Willebrand Factor (vWF) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Von Willebrand Factor (vWF). Target Synonyms: F8VWF; VWD; von Willebrand antigen 2. Accession Number: Q8CIZ8. Gene ID: 22371. Expression Region: Asp1498~Val1665. Tag Info: N-terminal His Tag. Theoretical MW: 20kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5784