Learn More
Biomatik Corporation Recombinant Von Willebrand Factor A Domain Containing Protein 2 (vWA2), 100UG
Supplier:  Biomatik Corporation RPU44374
Encompass_Preferred
Recombinant Human Von Willebrand Factor A Domain Containing Protein 2 (vWA2) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Von Willebrand Factor A Domain Containing Protein 2 (vWA2). Target Synonyms: AMACO; VWFA2; CCSP-2; A Domain-Containing Protein Similar To Matrilin And Collagen; Colon cancer secreted protein 2. Accession Number: Q5GFL6. Gene ID: 340706. Expression Region: Ser457~Gly708. Tag Info: N-terminal His Tag. Theoretical MW: 30/28kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5785