Learn More
Biomatik Corporation Recombinant Z-DNA Binding Protein 1 (ZBP1), 100UG
Catalog No. 501505788
Encompass_Preferred
Recombinant Human Z-DNA Binding Protein 1 (ZBP1) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Z-DNA Binding Protein 1 (ZBP1). Target Synonyms: DLM1; DAI; DNA-Dependent Activator Of IRFs; Tumor stroma and activated macrophage protein DLM-1. Accession Number: Q9H171. Gene ID: 81030. Expression Region: Arg10~Pro167. Tag Info: N-terminal His Tag. Theoretical MW: 21kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5788 Supplier Biomatik Corporation Supplier No. RPU44394
Recombinant Human Z-DNA Binding Protein 1 (ZBP1) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Z-DNA Binding Protein 1 (ZBP1). Target Synonyms: DLM1; DAI; DNA-Dependent Activator Of IRFs; Tumor stroma and activated macrophage protein DLM-1. Accession Number: Q9H171. Gene ID: 81030. Expression Region: Arg10~Pro167. Tag Info: N-terminal His Tag. Theoretical MW: 21kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.