Learn More
Biomatik Corporation Recombinant Human Stanniocalcin-1 (STC1), partial, 100UG
Supplier:  Biomatik Corporation RPC20193
Encompass_Preferred
Recombinant Human Stanniocalcin-1 (STC1), partial is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: STC1. Target Synonyms: Stanniocalcin 1; Stanniocalcin; Stanniocalcin-1; STC; STC-1; Stc1; STC1_HUMAN. Accession Number: P52823. Expression Region: 39~247aa. Tag Info: N-Terminal Gst-Tagged. Theoretical MW: 50.6kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4655