Learn More
Biomatik Corporation Native Hemoglobin (HB), 100UG
Supplier:  Biomatik Corporation RPU50181
Encompass_Preferred
Native Rat Hemoglobin (HB) is a purified Natural Protein. Purity: >95% by SDS-PAGE. Host: Rat. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Rat (Rattus norvegicus). Target Name: Hemoglobin (HB). Target Synonyms: Hgb; Haemoglobin; Heterotetramer(αβ)2. Accession Number: P01946; P02091; P11517. Theoretical MW: 14/16kDa. Buffer: PBS, pH7.4, containing 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5791